Learn More
Thermo Scientific™ CaptureSelect™ TSH Ligand Leakage ELISA
Sfoglia Tutto Thermo Scientific ProdottiDescrizione
CaptureSelect TSH Ligand Leakage ELISA is designed to detect possible leached ligand of thyroid-stimulating hormone (TSH) purified using CaptureSelect TSH Affinity Matrix. The ligand, a 14-kD recombinant protein, is covalently coupled for high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein.
This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect TSH Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.
Specifica
Specifica
| Content And Storage | Store at -5°C to -30°C. |
| Shipping Condition | Dry Ice |
| Product Line | CaptureSelect |
| Quantità | 1 Assay |
| Tipo | ELISA Kit |
| Formato | Kit |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.