missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase IV Monoclonal Antibody (310413), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA524028
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
In direct ELISAs and Western blots, less than 10% cross-reactivity with recombinant human (rh) CA1, rhCA2, and recombinant mouse CA4 is observed. This antibody does not cross-react with rhCA3, 8, 9, 10, 12, 13, or 14. Reconstitute at 0.5 mg/mL in sterile PBS.
Carbonic Anhydrase IV Monoclonal antibody specifically detects Carbonic Anhydrase IV in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blot.Specifica
Carbonic Anhydrase IV | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P22748 | |
CA4 | |
Mouse myeloma cell line NS0-derived recombinant human Carbonic AnhydraseIV/CA4 Ala19-Lys283 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
310413 | |
Unconjugated | |
CA4 | |
AW456718; CA IV; CA4; CAIV; CA-IV; Car4; Carbonate dehydratase IV; carbonic anhydrase 4; Carbonic anhydrase IV; carbonic dehydratase IV; RP17 | |
Mouse | |
Protein A/G | |
RUO | |
762 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto