missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase IV Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA578900
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL.
Carbonic anhydrases are a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. They participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. They show extensive diversity in tissue distribution and in their subcellular localization. This gene encodes a glycosylphosphatidyl-inositol-anchored membrane isozyme expressed on the luminal surfaces of pulmonary capillaries and proximal renal tubules. Its exact function is not known, however, it may have a role in inherited renal abnormalities of bicarbonate transport.Specifica
Carbonic Anhydrase IV | |
Polyclonal | |
Unconjugated | |
CA4 | |
AW456718; CA IV; CA4; CAIV; CA-IV; Car4; Carbonate dehydratase IV; carbonic anhydrase 4; Carbonic anhydrase IV; carbonic dehydratase IV; RP17 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12351, 29242, 762 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 4MG trehalose and 0.05MG sodium azide | |
P22748, P48284, Q64444 | |
CA4, Car4 | |
E.coli-derived human CA IV recombinant protein (Position: A19-S284). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto