Learn More
Emma Safety Footwear CAS Safety Shoes
Washable microfiber uppers are treated to repel all kinds of dirt, germs and bacteria. A highly breathable, comfortable shoe for all day wear. Energy returning soles prevent fatigue and keep the wearer fresh, alert, foot sure and steady even in the event of a spillage.
Marca: Emma Safety Footwear MM52054035
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- S2-Safety shoe for wet working conditions
- GripForce™ Easy Twist Outsole
- Microfiber construction
- Chrome-free lining
- Hydro-Tec™ Sanitized Silver
Specifica
EN ISO 20345:2011 | |
White | |
Hydro-Tec™ Sanitized Silver | |
35 |
Laces | |
CAS | |
Microfiber | |
Steel |