missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD178 Mouse anti-Human, BUV737, Clone: NOK-1, BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 748972
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
CD178 Monoclonal antibody specifically detects CD178 in Human samples. It is validated for Flow CytometrySpecifica
CD178 | |
Monoclonal | |
Brilliant Ultraviolet 737 | |
Fas ligand; FASL; FASLG; CD95 ligand; CD95L; CD95-L; TNFSF6; APTL; APT1LG1 | |
Mouse T lymphoma cells (L5178Y) expressing human FasL | |
RUO | |
Human | |
IgG1 |
Flow Cytometry | |
NOK-1 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV737 under optimal conditions that minimize unconjugated dye and antibody. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto