missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD63 Rabbit anti-Human, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech CL594-25682-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
CD63 Polyclonal antibody apecifically detetects CD63 antigen in Human Samples. It is validated for Flow Cytometry.Specifica
CD63 | |
Polyclonal | |
CoraLite 594 | |
BC002349 | |
Rabbit | |
Antigen affinity purification | |
Primary | |
Human | |
Liquid |
Flow Cytometry | |
1000 ug/mL | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
CD63 | |
CD63 fusion protein Ag19690 | |
20 μL | |
967 | |
Store at -20°C. Avoid exposure to light. | |
IgG |