Learn More
CDC42SE1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA555669
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: LGCCVVEKPQ PKKKRRRIDR TMIGEPMNFV HLTHIGSGEM GAGDGLAMTG AVQEQMRSKG NR Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
CDC42SE1 Polyclonal antibody specifically detects CDC42SE1 in Human samples. It is validated for Immunohistochemistry.Specifica
CDC42SE1 | |
Polyclonal | |
Unconjugated | |
CDC42SE1 | |
1300002M12Rik; CDC42 small effector 1; CDC42 small effector protein 1; CDC42-binding protein SCIP1; CDC42SE1; SCIP1; signaling molecule SPEC1 beta; small effector of CDC42 protein 1; small protein effector 1 of Cdc42; SPEC1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56882 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NRR8 | |
CDC42SE1 | |
Recombinant protein corresponding to Human CDC42SE1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only