Learn More
CDHR2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA553540
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: DVSLDYETQP VFNLTVSAEN PDPQGGETIV DVCVNVKDVN DNPPTLDVAS LRGIRVAENG SQHGQVAVVV ASDVDTSAQL EIQLVNILCT KAGVDVGSLC WGWFSVAANG SVYINQSKAI DYEACDLVTL VVRACDLATD P Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 74%.
CDHR2 Polyclonal antibody specifically detects CDHR2 in Human samples. It is validated for Immunohistochemistry.Specifica
CDHR2 | |
Polyclonal | |
Unconjugated | |
CDHR2 | |
Cadherin; cadherin related family member 2; cadherin-related family member 2; CDHR2; Gm624; PCDH24; PCLKC; PC-LKC; protocadherin 24; protocadherin LKC; Protocadherin-24 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54825 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BYE9 | |
CDHR2 | |
Recombinant protein corresponding to Human CDHR2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only