missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK5RAP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA538871
Dettagli aggiuntivi : Peso : 0.00900kg
Descrizione
This gene encodes a regulator of CDK5 (cyclin-dependent kinase 5) activity. The protein encoded by this gene is localized to the centrosome and Golgi complex, interacts with CDK5R1 and pericentrin (PCNT), plays a role in centriole engagement and microtubule nucleation, and has been linked to primary microcephaly and Alzheimer′s disease. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Jan 2013].Specifica
CDK5RAP2 | |
Polyclonal | |
Unconjugated | |
CDK5RAP2 | |
CEP215, KIAA1633 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55755 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q96SN8 | |
CDK5RAP2 | |
A synthetic peptide derived from the N-terminal region of human CDK5RAP2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |