Learn More
CDKAL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA529077
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Recommended positive controls: Juarkat, Raji, K562, NIH-3T3. Predicted reactivity: Mouse (98%), Zebrafish (84%), Xenopus laevis (90%), Dog (96%), Cat (97%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a member of the methylthiotransferase family. The function of this gene is not known. Genome-wide association studies have linked single nucleotide polymorphisms in an intron of this gene with susceptibility to type 2 diabetes.Specifica
CDKAL1 | |
Polyclonal | |
Unconjugated | |
Cdkal1 | |
1190005B03Rik; 6620401C13Rik; CDK5 regulatory subunit associated protein 1 like 1; CDK5 regulatory subunit associated protein 1-like 1; CDK5 regulatory subunit-associated protein 1-like 1; CDKAL; CDKAL1; Threonylcarbamoyladenosine tRNA methylthiotransferase; tRNA-t(6)A37 methylthiotransferase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54901, 68916 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q5VV42, Q91WE6 | |
Cdkal1 | |
Recombinant fragment corresponding to a region within amino acids 3 and 282 of Human CDKAL1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |