missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDNF Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marca: Invitrogen PA547708
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
In direct ELISAs, approximately 30% cross-reactivity with recombinant human CDNF is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
CDNF Polyclonal antibody specifically detects CDNF in Mouse samples. It is validated for Immunohistochemistry (Frozen), Western Blot.Specifica
CDNF | |
Polyclonal | |
Unconjugated | |
Cdnf | |
9330140G23; arginine-rich protein mutated in early stage tumors-like 1; arginine-rich, mutated in early stage tumors-like 1; ARMETL1; ARMET-like protein 1; Cdnf; Cerebral dopamine neurotrophic factor; conserved dopamine neurotrophic factor | |
Goat | |
Antigen affinity chromatography | |
RUO | |
227526 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Frozen), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q8CC36 | |
Cdnf | |
Mouse myeloma cell line NS0-derived recombinant mouse CDNF Leu27-Leu187 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |