Learn More
Advantec Mfs™ Cellulose Ester Membrane
MCE membranes are popular for many applications in the microbiological examination of water, wastewater, pharmaceuticals, food and beverages, and air monitoring. They offer high recovery rates and superior flow rates.
Marca: Advantec Mfs™ A045F047A
Dettagli aggiuntivi : CAS : 9004-70-0 Peso : 0.01000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II
Descrizione
- Mixed Cellulose Esters (MCE)
- High recovery rates and superior flow rates
- Contrasting gridlines facilitate counting colonies and are tested to assure freedom from grid line inhibition
- EtO sterilised
- Individually packaged
- 47 mm Diameter
Specifica
White | |
Gridded | |
Cellulose Ester | |
0.45 μm | |
For capturing and culturing microorganisms using the MF Technique in the Water and Wastewater Industry | |
EtO | |
1.9 in. |
White membrane filter with pad | |
Disc | |
Cellulose Ester | |
Sterile | |
100 Pk. | |
Membrane Filter | |
47 mm |