Learn More
LABPLAS Cellulose Sani-Stick with Buffered Peptone Water
Cellulose sponge with stick for sampling surfaces, with buffer to neutralize disinfectant.
Marca: LABPLAS KSS-67310-BPW
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Cellulose Sani-Stick
- Saturated with buffered peptone water
- Dimensions: 160 x 229 mm
- Holds: 600 mL
- Option to include gloves with kit
Specifica
Hygiene Sponge Sampling Kit | |
Neutralizers | |
Sterile | |
None |