missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CELSR3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA538869
Dettagli aggiuntivi : Peso : 0.00900kg
Descrizione
CELSR3 is a member of the flamingo subfamily, part of the cadherin superfamily. The flamingo subfamily consists of nonclassic-type cadherins; a subpopulation that does not interact with catenins. The flamingo cadherins are located at the plasma membrane and have nine cadherin domains, seven epidermal growth factor-like repeats and two laminin A G-type repeats in their ectodomain. They also have seven transmembrane domains, a characteristic unique to this subfamily. It is postulated that these proteins are receptors involved in contact-mediated communication, with cadherin domains acting as homophilic binding regions and the EGF-like domains involved in cell adhesion and receptor-ligand interactions. The specific function of this particular member has not been determined.Specifica
CELSR3 | |
Polyclonal | |
Unconjugated | |
CELSR3 | |
CELSR3, Anchor protein, Cadherin family member 11, CDHF11, EGF-like protein 1, EGF-like-domain, multiple 1, Epidermal growth factor-like 1, Flamingo homolog 1, FMI1, KIAA0812, MEGF2, Megf3, HFMI1, Multiple EGF-like domains 2, RESDA1, EGFL1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1951 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q9NYQ7 | |
CELSR3 | |
A synthetic peptide derived from the internal region of human CELSR3 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |