Learn More
CEP162 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA556382
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: NISTMESDLP TVEELMKPIR IDSFGISGFD LQPVSSEKVA ERKETEFFSS LPLKMNPNIL SQDSQHVNLF FDKNDENVIL QKTTNESMEN SCPQVTEVTA TEE Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 57%.
CEP162 Polyclonal antibody specifically detects CEP162 in Human samples. It is validated for Immunohistochemistry.Specifica
CEP162 | |
Polyclonal | |
Unconjugated | |
CEP162 | |
C6orf84; centrosomal protein 162; centrosomal protein 162kDa; centrosomal protein of 162 kDa; Cep162; KIAA1009; protein QN1 homolog; QN1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22832 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5TB80 | |
CEP162 | |
Recombinant protein corresponding to Human CEP162 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only