Learn More
Koettermann™ Ceramic Sink
Click to view available options
:
Includes: Marine edge, 1 bowl, left
Descrizione
- Marine edge on all sides to contain spills
- One sink bowl, left
- Baltic blue color
Specifica
Specifica
| Tipo | Laboratory Sink |
| Include | Marine edge, 1 bowl, left |
| Larghezza (sistema metrico) | 1200 mm |
| Profondità (sistema metrico) | 600 mm |