Learn More
CERK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA532706
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey, Panda (100%); Gorilla, Dog, Elephant, Horse (93%); Mouse, Rat, Hamster (86%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
CERK converts ceramide to ceramide 1-phosphate (C1P), a sphingolipid metabolite. Both CERK and C1P have been implicated in various cellular processes, including proliferation, apoptosis, phagocytosis, and inflammation (Kim et al., 2006 ).Specifica
CERK | |
Polyclonal | |
Unconjugated | |
CERK | |
acylsphingosine kinase; AI848610; Ceramide kinase; Cerk; CERK antibody; D330016D08Rik; dA59H18.2; dA59H18.3; hCERK; KIAA1646; Lipid kinase 4; lipid kinase LK4; LK4; mCERK; mKIAA1646 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64781 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q8TCT0 | |
CERK | |
Synthetic 14 amino acid peptide from internal region of human CERK. | |
50 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |