Learn More
CF106 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA556949
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: MEGMDVDLDP ELMQKFSCLG TTDKDVLISE FQRLLGFQLN PAGCAFFLDM TNWNLQAAIG AYYDFESPNI SVPSMS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.
Negative regulator of innate antiviral response. Blocks IRF3-dependent cytokine production such as IFNA, IFNB and TNF (PubMed:29802199). Interacts with IRF3 and inhibits IRF3 recruitment to type I IFN promoter sequences while also reducing nuclear levels of the coactivators EP300 and CREBBP (PubMed:29802199). [UniProt]Specifica
CF106 | |
Polyclonal | |
Unconjugated | |
C6orf106 | |
AU020189; C030010A15; C6orf106; C80239; chromosome 6 open reading frame 106; D17Wsu92e; dJ391O22.4; DNA segment, Chr 17, Wayne State University 92, expressed; FP852; Ilrun; Inflammation and lipid regulator with UBA-like and NBR1-like domains protein; Protein ILRUN; uncharacterized protein C6orf106; uncharacterized protein C6orf106 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64771 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H6K1 | |
C6orf106 | |
Recombinant protein corresponding to Human CF106 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only