Learn More
CHAC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA531762
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Recommended positive controls: HeLa(4ug/ml TM_12hr, 4ug/ml TM_24hr)(30ug). Predicted reactivity: Mouse (94%), Rat (96%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Pro-apoptotic component of the unfolded protein response pathway. May mediate the pro-apoptotic effects of the ATF4-ATF3-DDIT3/CHOP cascade.Specifica
CHAC1 | |
Polyclonal | |
Unconjugated | |
CHAC1 | |
1810008K03Rik; Blocks Notch protein; botch; cation transport regulator homolog 1; cation transport regulator-like protein 1; ChaC glutathione specific gamma-glutamylcyclotransferase 1; ChaC glutathione-specific gamma-glutamylcyclotransferase 1; ChaC, cation transport regulator 1; ChaC, cation transport regulator homolog 1; ChaC, cation transport regulator-like 1; Chac1; Gamma-GCG 1; gamma-GCT acting on glutathione homolog 1; glutathione-specific gamma-glutamylcyclotransferase 1; Neuroprotective protein 7; Npg7; RGD1307153 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79094 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9BUX1 | |
CHAC1 | |
Recombinant fragment corresponding to a region within amino acids 38 and 254 of Human CHAC1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |