Learn More
OHAUS™ Chain Clamp
Sfoglia Tutto Ohaus Prodotti
Click to view available options
Size:
Small
Large
Materiale:
Nickel-plated
Stainless Steel
Lunghezza (sistema metrico) braccio:
127 mm
305 mm
Dimensione della confezione:
Pezzo
Descrizione
- Corrosion-resistant stainless steel clamps provide excellent chemical resistance, durability, and can be autoclaved. Nickel-plated zinc clamps offer high-tensile strength at an economical price.
- Prevent accidents in the lab! Versatile multi-purpose LabJaws clamps securely hold glassware and irregularly-shaped objects. Select from a variety of grip sizes, prong styles and adjustment types.
- Choose from a variety of specialized LabJaws clamps with adjustable features to provide an exact fit for lab ware such as thermometers, thermocouples, columns, funnels, and round bottom flasks.
Specifica
Specifica
| Tipo | Chain Clamp |
| Materiale | Stainless Steel |
| Descrizione | Clamp, Specialty, Chain CLS-CHAINSL |
| Diametro (sistema metrico) | 13 mm |
| Lunghezza (sistema imperiale) | 8.11 in. |
| Lunghezza (sistema imperiale) braccio | 5.000 in. |
| Lunghezza (sistema metrico) | 206.00 mm |
| Lunghezza (sistema metrico) braccio | 127 mm |
| Size | Large |
| Dimensioni (sistema metrico) impugnatura | 35 to 280 mm |