Learn More
Nabertherm™ Chamber Furnace LH 1200 Series with C540 Controller
Sfoglia Tutto Nabertherm Prodotti
Click to view available options
Capacità (sistema metrico):
15 L
30 L
60 L
120 L
216 L
Dimensione della confezione:
Pezzo
Descrizione
- Maximum Temperature: 1200°C
- Dual shell housing with rear ventilation, provides for low shell temperatures
- Five-sided heating for very good temperature uniformity
- Heating elements on support tubes ensure free heat radiation and a long service life
- Controller mounted on furnace door and removable for comfortable operation
- Protection of bottom heating and flat stacking surface provided by embedded SiC plate in the floor
- Multi-layered, fiber-free insulation of light refractory bricks and special backup insulation
- Door with brick-on-brick seal, hand fitted
- Short heating times due to high installed power
- Side vent with bypass connection for exhaust pipe
- Self-supporting arch for high stability and greatest possible protection against dust
- Quick lock on door
- Freely adjustable air slide intake in furnace floor
- Stand included
- Defined application within the constraints of the operating instructions
Specifica
Specifica
| Tipo di unità di controllo | C540 |
| Capacità (sistema metrico) | 120 L |
| Temperatura (sistema metrico) | 1200°C |
| Dimensioni interne (L x P x A) | 500 x 500 x 500 mm |
| Dimensioni esterne (L x P x A) | 1170 x 880 x 1465 mm |
| Peso (sistema metrico) | 410 kg |