Learn More
Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers
Record seven days of temperature performance at temperatures to -150° C.
Marca: Thermo Scientific™ FCRP500LE
Dettagli aggiuntivi : Peso : 14.00000kg
Description
I registratori grafici per congelatori a bassissima temperatura Thermo Scientific™ consentono di registrare l'andamento settimanale delle temperature fino a -150°C. I registratori utilizzano carta per grafici 6 in. (15 cm) e possono essere installati in loco o in fabbrica.
Field-installed inkless chart recorderPrestations de service
Warranty varies by country, please contact us for details
Spécification
Ink Chart Recorder, Field Installed | |
Chart Recorder | |
Revco RLE, Forma 89000, TLE Series, HLE Series (500 models only) | |
Recorder |