missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHCHD2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA565727
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Immunogen sequence: PRGSRSRTSRM APPASRAPQM RAAPRPA Highest antigen sequence identity to the following orthologs - mouse 89%, rat 86%.
Transcription factor. Binds to the oxygen responsive element of COX4I2 and activates its transcription under hypoxia conditions (4% oxygen), as well as normoxia conditions (20% oxygen).Specifica
CHCHD2 | |
Polyclonal | |
Unconjugated | |
CHCHD2 | |
16.7kD protein; AAG10; Aging-associated gene 10 protein; AL033347; C7orf17; CHCHD2; coiled-coil-helix-coiled-coil-helix domain containing 2; coiled-coil-helix-coiled-coil-helix domain-containing protein 2; coiled-coil-helix-coiled-coil-helix domain-containing protein 2, mitochondrial; ethanol induced 6; Etohi6; HCV NS2 trans-regulated protein; mitochondria nuclear retrograde regulator 1; MNRR1; NS2TP; PARK22 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51142 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y6H1 | |
CHCHD2 | |
Recombinant Human CHCHD2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only