Learn More
Lovibond™ CHECKIT™ Comparator Test Kits: Total Hardness HR
Accurate test kit for routine water analysis
Marca: Lovibond™ 414240
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
Routine water analysis | |
100 | |
Total Hardness Mini Kit |
30mL calibrated sample container, 30mL calibrated sample container, total hardness tablets, instructions, storage case | |
5 to 500mg/L |