Learn More
Lovibond™ Checkit™ Comparators 2-in-1 Test Kits
Click to view available options
Range:
6.5 to 8.4 pH, 0 to 1 mg/L Cl2
6.5 to 8.4 pH, 0 to 1 mg/L Cu
6.5 to 8.4 pH, 0 to 4 mg/L Cl2
6.5 to 8.4 pH, 0 to 5 mg/L Br
6.5 to 8.4 pH, 0.1 to 2 mg/L Cl2
Descrizione:
pH Value
Copper
Copper
pH Value
Bromine
Bromine
pH value
Chlorine
Chlorine
pH value
Chlorine
Chlorine
Dimensione della confezione:
Pezzo
Descrizione
Perform water analysis quickly and easily. Lovibond Checkit™ Comparitors 2-in-1 Test Kits are highly accurate and incorporate a user-friendly colorimetric scale. Each kit indicates a combination of pH and another measure.
- Easy operation and exact reagent dosing
- Tablet reagents with a minimm guaranteed shelf life of 5/10 years
- Rotate continuous gradient color wheel to obtain a color match with the reacted sample
- Printed continuous gradient color discs provide accurate measurements without "exrapolating" between discrete color blocks
Specifica
Specifica
| Descrizione | pH value Chlorine |
| Range | 6.5 to 8.4 pH, 0.1 to 2 mg/L Cl2 |
| Metodo test | Colorimetric |
| Tipo | 2 in 1 Test Kit |