Learn More
Lovibond™ Checkit™ Comparitor Test Kits
Click to view available options
Range:
0 to 0.5 mg/L
0 to 2.5 mg/L
0.01 to 0.2 mg/L
Descrizione:
Chlorine dioxide
DEHA
Phosphate, powder pack
Dimensione della confezione:
Pezzo
Descrizione
Perform water analysis quickly and easily. Lovibond Checkit™ Comparitor Test Kits are highly accurate and incorporate a user-friendly colorimetric scale. Each kit indicates a combination of pH and another measure.
- Easy operation and exact reagent dosing
- Tablet reagents with a minimm guaranteed shelf life of 5/10 years
- Rotate continuous gradient color wheel to obtain a color match with the reacted sample
- Printed continuous gradient color discs provide accurate measurements without "exrapolating" between discrete color blocks
Specifica
Specifica
| Descrizione | Phosphate, powder pack |
| Range | 0 to 2.5 mg/L |
| Metodo test | Colorimetric |
| Tipo | Water test kit |