Learn More
DiscoverX ChemiScreen™ BB3 Bombesin Receptor Membrane Preparation
The ChemiScreen™ BB3 Bombesin Receptor Membrane Preparation allows measurement of ligand binding to BRS3 GPCR via radioligand binding assays. Membrane preps contain 1mL of 1mg/mL frozen membranes.
Marca: DiscoverX HTS160M
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The ChemiScreen™ BB3 Bombesin Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of BRS3 surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1mL of 1mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression.Specifica
BB3 | |
Membrane Preps | |
1 Vial (200 units) | |
BRS3 |