Learn More
DiscoverX ChemiScreen™ BB3 Bombesin Receptor Stable Cell Line
The ChemiScreen™ BB3 Bombesin Receptor Stable Cell Line measures BRS3 (GPCR) activation via an increase in intracellular calcium. This cell line ships as two vials of cryopreserved cells.
Marca: DiscoverX HTS160C
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The ChemiScreen™ BB3 Bombesin Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. bombesin) based activation of BRS3 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses BRS3 alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of BRS3 , the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells.Specifica
Calcium Flux | |
Cell Lines | |
2 Vials | |
NM_001727 |
Chem-1 | |
[D-Tyr6, β-Ala11, Phe13, Nle14]-Bombesin(6-14) | |
BRS3 |