Learn More
DiscoverX ChemiScreen™ Y4 Neuropeptide Y Receptor Membrane Preparation
The ChemiScreen™ Y4 Neuropeptide Y Receptor Membrane Preparation allows measurement of ligand binding to NPY4R GPCR via radioligand binding assays. Membrane preps contain 1mL of 1mg/mL frozen membranes.
Marca: DiscoverX HTS204M
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The ChemiScreen™ Y4 Neuropeptide Y Receptor Membrane Preparation is derived from proprietary stable recombinant cells to ensure high levels of NPY4R surface expression. ChemiScreen membrane preps can be used to screen for ligand binding affinities using a competitive radio-labeled ligand approach. Increased binding affinities will lead to higher radioactive signal. Membrane preps contain 1mL of 1mg/mL frozen membranes derived from a stably modified clonal cell line which was chosen for optimal receptor expression.Specifica
Y4 | |
Membrane Preps | |
1 Vial (200 units) | |
NPY4R |