Learn More
Chitotriosidase Monoclonal Antibody (1D9G2,1D9G4,2B4G3), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA515482
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
MA5-15482 targets CHIT1 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15482 immunogen is purified recombinant fragment of CHIT1 (aa22-137) expressed in E. Coli.<br>.
Chitotriosidase is secreted by activated human macrophages and is markedly elevated in plasma of Gaucher disease patients. The expression of chitotriosidase occurs only at a late stage of differentiation of monocytes to activated macrophages in culture. Human macrophages can synthesize a functional chitotriosidase, a highly conserved enzyme with a strongly regulated expression. This enzyme may play a role in the degradation of chitin-containing pathogens.Specifica
Chitotriosidase | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
Q13231 | |
CHIT1 | |
Purified recombinant fragment of CHIT1 (aa22-137) expressed in E. Coli. | |
RUO | |
1118 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
1D9G2,1D9G4,2B4G3, 1D9G4, 2B4G3 | |
Unconjugated | |
CHIT1 | |
2300002L19Rik; CHI3; CHIT; Chit1; CHITD; chitinase 1; chitinase 1 (chitotriosidase); chitinase-1; Chitotriosidase; Chitotriosidase-1; EC 3.2.1.14; FLJ00314; MGC125322; plasma methylumbelliferyl tetra-N-acetylchitotetraoside hydrolase | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |