Learn More
Berkshire Choice™ SuperSorb
Choice™ SuperSorb is a 2-ply, high absorbency, ultrasonically sealed edge, cleanroom laundered wiper.
Marca: Berkshire CHSS0414
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- 100% continuous filament knit polyester
- Two-ply pinsonic construction with unique diamond pattern
- Ultrasonically sealed edge for reduced fiber contamination
- Laundered and packaged in ISO Class 4 (Class 10) cleanroom
- Critically low particles, fibers, ions and extractables
- High abrasion resistance
- Chemically compatible with IPA, acetone and other solvents
- Pinsonic design adds surface texture that aids in stubborn residue removal
Specifica
Surface Wiper | |
10 cm | |
10 cm | |
Autoclavable | |
2-ply |
4 in. | |
4 in. | |
Polyester | |
Single-use | |
Non-sterile |