Learn More
Thermo Scientific Chemicals Cholecystokinin, CCK Octapeptide (26-33)
For cellular and molecular biology applications
85.93€ - 131.66€
Specifica
Metodo di purificazione HPLC
Nome comune CCK 8
Content And Storage -30°C to -10°C
Formato Solid
Peso molecolare 1143.27
Visualizza altre specifiche
Prodotti 2
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
15418119
Vedere i documenti
Thermo Scientific Chemicals
0.5 mg
85.93€
0.50mg
15428119
Vedere i documenti
Thermo Scientific Chemicals
1 mg
131.66€
1mg
Descrizione
Descrizione
Cholecystokinin, CCK Octapeptide (26-33) is used in the biological studies in the preparation of opioid receptors agonistic and CCK receptor antagonistic activity. It plays a role in satiety.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
HPLC
-30°C to -10°C
1143.27
RUO - research use only
CCK 8
Solid
Cholecystokinin, CCK Octapeptide (26-33)
Asp-Tyr(SO3H)-Met-Gly-Trp-Met-Asp-Phe-NH2