Learn More
Chromogranin B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA110839
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
PA1-10839 detects Chromogranin B from human, rat, and mouse samples. PA1-10839 has been successfully used in Western blot and immunohistochemistry applications. The PA1-10839 immunogen is a synthetic peptide, conjugated to ovalbumin, corresponding to the 15 most C-terminal residues of chromogranin B.
Chromogranin B is a tyrosine sulfated secretory protein found in a wide variety of peptidergic endocrine cells. Chromogranin functions as a neuroendocrine secretory granule protein which may be the precursor for other biologically active peptides.Specifica
Chromogranin B | |
Polyclonal | |
Unconjugated | |
Chgb | |
CCB peptide; CCB peptide long form; CCB peptide short form; CgB; CHGB; chromogranin B; chromogranin B (secretogranin 1); Chromogranin B, parathyroid secretory protein; chromogranin-B; GAWK peptide; Glucagonoma peptide; PE-11; SCG1; Scg-1; secretogranin B; secretogranin I; Secretogranin-1; secretogranin-I; SgI | |
Rabbit | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Conc. Not Determined | |
whole serum with no preservative | |
O35314, P05060, P16014 | |
Chgb | |
Synthetic peptide, conjugated to ovalbumin, corresponding to the 15 most C-terminal residues of chromogranin B. | |
RUO | |
1114, 12653, 24259 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |