Learn More
CHST4 Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Marca: Invitrogen PA547752
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant human (rh) CHST4 is observed and less than 5% cross-reactivity with recombinant mouse CHST5 and rhCHST6 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
CHST4 Polyclonal antibody specifically detects CHST4 in Mouse samples. It is validated for Neutralization, Western Blot.Specifica
CHST4 | |
Polyclonal | |
Unconjugated | |
Chst4 | |
carbohydrate (chondroitin 6/keratan) sulfotransferase 4; carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 4; carbohydrate sulfotransferase 4; Chst4; galactose/N-acetylglucosamine/N-acetylgalactosamine 6-O-sulfotransferase 3; galactose/N-acetylglucosamine/N-acetylglucosamine; galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 3; GlcNAc6ST2; GlcNAc6ST-2; Gn6st-2; Gst3; GST-3; HECGLCNAC6ST; HEC-GlcNAc6ST; high endothelial cell GlcNAC-6-sulphotransferase; high endothelial cells N-acetylglucosamine 6-O-sulfotransferase; L-selectin ligand sulfotransferase; LSST; N-acetylglucosamine 6-O-sulfotransferase 2; sulfo6 | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
26887 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9R1I1 | |
Chst4 | |
CHO-derived recombinant mouse Carbohydrate Sulfotransferase 4/CHST4 Arg30-Gly388 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |