Learn More
Thermo Scientific™ Unità di controllo Cimarec™ i 40C 100-240V
Receive higher speeds and more power settings than standard controllers by using Thermo Scientific™ Cimarec™ i 40C Controllers.
Marca: Thermo Scientific™ 50090774
Dettagli aggiuntivi : Peso : 2.27700kg
Descrizione
Receive higher speeds and more power settings than standard controllers by using Thermo Scientific™ Cimarec™ i 40C Controllers. Cimarec™ i 40C Controllers are included with the Maxi and Telesystems Stirrers.
Features:- Power output booster to 40w to handle more demanding stirring tasks
- 10 power settings to decrease power consumption and lower heat output
- Single-handed operation with straightforward turn and press adjustment wheel
- Three individual program store and recall keys for rapid start-up
- Last used settings stored in memory
- Rocking/shake mode reverses stir bar direction for a gentle back and forth stirring motion
- RS-232 interface for data transfer and PC control
Ordering Information:
Order as replacement controller only.
Specifica
Controller 40C | |
9.5 cm | |
16.5 cm | |
40 w | |
±1 | |
Da 0° a 40 a 80% di umidità relativa °C | |
6.1 in. | |
±1 | |
100 RPM to 2000 RPM |
3.7 in. | |
6.4 in. | |
Da 5 sec. a 60 min. | |
Da 100 a 2000 rpm | |
Da 5 sec. a 60 min. | |
Stirrer Controller | |
15.5 cm | |
100/240 V | |
Da 10 a 100 (10 livelli) |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.