Learn More
Tocris Bioscience™ CINPA 1
Potent CAR antagonist
Marca: Tocris Bioscience™ 5605/10
Dettagli aggiuntivi : CAS : 102636-74-8 Peso : 0.00970kg
Descrizione
CINPA 1 is a potent CAR antagonist (IC50 = 70 nM). Exhibits >90-fold selectivity for CAR over pregnane X receptor (PXR). Also selective over a panel of other nuclear receptors including FXR, LXR, PPARγ and RXR.Specifica
Ethyl [5-[(diethylamino)acetyl]-10,11-dihydro-5H-dibenz[b,f]azepin-3-yl]carbamate | |
102636-74-8 | |
10 mg |