Learn More
Labconco™ Circulation Tubing Kit
Connects AtmoSense Oxygen and Moisture Monitors in-line between the Protector Controlled Atmosphere or Combination Glove Box or the Precise Controlled Atmosphere Glove Box and the AtmosPure Re-Gen Gas Purifier.
Marca: Labconco™ 5441808
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Two 2cm (0.75 in.) I. D. flexible neoprene tubes
- Brass fittings 2cm (0.75 in.) O.D. on each end of tubing
- Includes two sections of tubing with fitting
Specifica
Tubing Kit |