Learn More
cis-1-Ethoxy-2-(tri-n-butylstannyl)ethylene, 94%, Thermo Scientific Chemicals
Catalog Number 15458077
Click to view available options
Quantità:
1 g
5 g
64724-29-4
C16H34OSn
361.157
MFCD07787391
WARKYKQCOXTIAO-UHFFFAOYSA-N
z-1-ethoxy-2-tributylstannyl ethene, tributyl z-2-ethoxyvinyl stannane, tributyl z-2-ethoxyethenyl stannane, tributyl-z-2-ethoxyethenyl stannane, z-tributyl 2-ethoxyvinyl stannane, cis-1-ethoxy-2-tri-n-butylstannyl ethylene, cis-tributyl 2-ethoxyethenyl stannane, tributyl-z-2-ethoxyvinyl stannane, z-1-ethoxy-2-tributylstannyl ethylene
5706773
tributyl-[(Z)-2-ethoxyethenyl]stannane
CCCC[Sn](CCCC)(CCCC)C=COCC
64724-29-4
C16H34OSn
361.157
MFCD07787391
WARKYKQCOXTIAO-UHFFFAOYSA-N
z-1-ethoxy-2-tributylstannyl ethene, tributyl z-2-ethoxyvinyl stannane, tributyl z-2-ethoxyethenyl stannane, tributyl-z-2-ethoxyethenyl stannane, z-tributyl 2-ethoxyvinyl stannane, cis-1-ethoxy-2-tri-n-butylstannyl ethylene, cis-tributyl 2-ethoxyethenyl stannane, tributyl-z-2-ethoxyvinyl stannane, z-1-ethoxy-2-tributylstannyl ethylene
5706773
tributyl-[(Z)-2-ethoxyethenyl]stannane
CCCC[Sn](CCCC)(CCCC)C=COCC
cis-1-Ethoxy-2-(tri-n-butylstannyl)ethylene is widely used in pharmaceutical research and as reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
cis-1-Ethoxy-2-(tri-n-butylstannyl)ethylene is widely used in pharmaceutical research and as reagent.
Solubility
Soluble in water.
Notes
cis-1-Ethoxy-2-(tri-n-butylstannyl)ethylene is to be stored in a cool, dry place and is to be sealed properly.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico cis-1-Ethoxy-2-(tri-n-butylstannyl)ethylene
Punto di fusione 84°C to 87°C
Densità 1.082
Quantità 1 g
Numero UN UN2788
Informazioni di solubilità Soluble in water.
Peso formulazione 361.15
Percent Purity 94%