missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-1-Ethoxy-2-(tri-n-butylstannyl)ethylene, 94%, Thermo Scientific Chemicals
Click to view available options
Quantità:
1 g
5 g
Descrizione
cis-1-Ethoxy-2-(tri-n-butylstannyl)ethylene is widely used in pharmaceutical research and as reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
cis-1-Ethoxy-2-(tri-n-butylstannyl)ethylene is widely used in pharmaceutical research and as reagent.
Solubility
Soluble in water.
Notes
cis-1-Ethoxy-2-(tri-n-butylstannyl)ethylene is to be stored in a cool, dry place and is to be sealed properly.
cis-1-Ethoxy-2-(tri-n-butylstannyl)ethylene is widely used in pharmaceutical research and as reagent.
Solubility
Soluble in water.
Notes
cis-1-Ethoxy-2-(tri-n-butylstannyl)ethylene is to be stored in a cool, dry place and is to be sealed properly.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifica
Materiale o nome chimico | cis-1-Ethoxy-2-(tri-n-butylstannyl)ethylene |
Punto di fusione | 84°C to 87°C |
Densità | 1.082 |
Quantità | 1 g |
Numero UN | UN2788 |
Informazioni di solubilità | Soluble in water. |
Peso formulazione | 361.15 |
Percent Purity | 94% |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'