Learn More
Citric acid, trisodium salt hydrate, 99.8%, for molecular biology, DNAse, RNAse and, Thermo Scientific Chemicals
Codice prodotto. 10076113
Click to view available options
Quantità:
250 g
1 kg
5 kg
Imballaggio:
Glass Bottle
Plastic bottle
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 114456-61-0
Formula molecolare C6H5Na3O7
Molecular Weight (g/mol) 258.07
Numero MDL MFCD00150032
InChI Key HRXKRNGNAMMEHJ-UHFFFAOYSA-K
Sinonimo trisodium citrate hydrate, sodium citrate tribasic hydrate, citric acid trisodium salt hydrate, sodium citrate, tribasic, sodium citrate monohydrate, c6h5o7.3na.h2o, sodium citrate tribasic hydrate, reagentplus r, trisodium 2-hydroxypropane-1,2,3-tricarboxylate hydrate
PubChem CID 21868242
SMILES [Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Specifica
Materiale o nome chimico Citric acid, trisodium salt hydrate
Nota nome For molecular biology
Absorbance 0.02 max. (0.1 M in water) at 260nm
Punto di fusione >300.0°C
Colore White
pH 7 to 9 (5% aq. soln.)
Perdita all'essiccatura 12% to 13%
Spettro a infrarossi Authentic
Intervallo percentuale saggio 0.998
Imballaggio Plastic bottle
Quantità 5 kg
Indice di Merck 14, 8602
Informazioni di solubilità Solubility in water: soluble.
IUPAC Name trisodium 2-hydroxypropane-1,2,3-tricarboxylate
Peso formulazione 258.08
Percent Purity 0.998
Grado Molecular Biology
Forma fisica Crystalline Powder or Crystals