Learn More
Honeywell™ Clearflow 3 Compressed Airline Filtration Unit, Portable Stand Model
Click to view available options
:
Airflow: 1600L/mn
Descrizione
- Floor model on portable stand with 2 CEJN outlets
- Supplies breathable air for up to 4 workers using supplied respirators, or 2 operators and auxiliary air-driven tools
- Uses coalescing filter and carbon absorber filter to remove oil vapor and odor
- High filtration capacity due to high flow system, high efficiency of coalescing filter, and high capacity of absorber carbon filter
- Easy use and easy maintenance due to new one-piece compact design
- Provides security to worker via accurate and reliable differential pressure indicator
- Filter
- Coalescing filter : 1000 hours with a filtration at 99,97% to 0.01 micron - Charcoal filter : 1000 hours (or when odors are detected inside respiratory device) Filters are compliant with ISO 8573 -1 Compressed Air - Part 1: Pollutants and purity classes) which specifies the quality class of compressed air for the presence of particles, water or oil, irrespective of the mode of generation of the compressed air)
Specifica
Specifica
| Applicazioni | Breathable air/air for air-driven tools |
| Portata | 1600 L/min. |
| Da utilizzare con (apparecchiatura) | Respirators/air-driven tools |
| Materiale | Aluminum body filters, plastic and brass regulators, polycarbonate bowl, nitrile seals, ABS cover |
| Pressione | 8 bar |