Learn More
Honeywell™ Clearways Visor CV83P/CG
Click to view available options
:
Trasparente
Descrizione
Includes
Visor only; headband is sold separately (12622733).
- Economical replaceable visors available in polycarbonate.
- Offers both strength and durability.
- Lightweight
- Comfortable for all-day wear
Specifica
Specifica
| Certificazione/Conformità | EC Category PPE: 2, EC Certificate: 0102, EC Attestation: 0086 |
| Descrizione | Visor |
| Lens Tint | Clear |
| Include | Visor only; headband is sold separately. |
| Lens Material | Policarbonato |