Learn More
Thermo Scientific™ CLINIConic™ Rotor Adapters
Codice prodotto. 10581845 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Capacità:
10 mL Round-bottom Tube
5 mL Round-bottom or Blood Collection Tube
7 mL Round-bottom or Blood Collection Tube
Dimensione della confezione:
Pezzo
Codice prodotto. 10581845
Marca: Thermo Scientific™ 11172596
Incorporate these rotor adapters with the CLINIConic fixed-angle rotor.
Incorporate Thermo Scientific™ CLINIConic™ Rotor Adapters with the CLINIConic fixed-angle rotor.
Ordering Information:
CLINIConic fixed-angle rotor body (75-003-623) is available separately. Contact your sales representative for more information.
TRUSTED_SUSTAINABILITY
Specifica
Tipo Rotor Tube Adapter
Tipo di provetta 7 mL Round Bottom, Blood Collection Tube
Capacità 7 mL Round-bottom or Blood Collection Tube