missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COL11A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA566160
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Immunogen sequence: DCDSSAPKAAQ AQEPQIDEYA PEDIIEYDYE YGEAEYKEAE SVTEGPTVTE ETIAQTEANI VDDFQEYNYG Highest antigen sequence identity to the following orthologs - mouse 85%, rat 86%.
Collagen Type XI is an 1806 amino acid protein belonging to the fibrillar collagen family. Collagen Type XI is thought to play an important role in fibrillogenesis by controlling lateral growth of collagen II fibrils. This protein forms trimers composed of three different chains: a 1(XI), a 2(XI), and a 3(XI). a 3(XI) is a post-translational modification of a 1(II). a 1(V) can also be found instead of a 3(XI). Collagen Type XI has three named isoforms (A,B,C) and additional isoforms seem to exist, stemming from alternative usage of exon IIA or exon IIB. Transcripts containing exon IIA or IIB are present in cartilage, but exon IIB is preferentially utilized in transcripts from tendon. Collagen Type XI contains a single collagen binding TSP N-terminal (TSPN) domain. Collagen Type XI is expressed in cartilage, placenta and some tumor or virally transformed cell lines. Isoform expression can be tissue specific. Defects in the COL11A gene are the cause of Stickler syndrome type 2 (STL2), or beaded vitreous type, due to the presence of irregularly thickened fiber bundles throughout vitreous cavity.
Specifica
| COL11A1 | |
| Polyclonal | |
| Unconjugated | |
| COL11A1 | |
| a1(XI) collagen; C530001D20Rik; cho; chondrodysplasia; CO11A1; COBA1; COL11A1; COLL6; Collagen alpha 1; collagen alpha-1(XI) chain; collagen type XI alpha 1 chain; collagen XI, alpha-1 polypeptide; collagen, type XI, alpha 1; pro-alpha1(XI) collagen; procollagen, type XI, alpha 1; STL2; STL3; XI chain precursor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1301 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P12107 | |
| COL11A1 | |
| Recombinant Human COL11A1. Recombinant protein control fragment (Product #RP-106845). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto