missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Columbia CNA Agar with Sheep Blood Medium
Click to view available options
Quantità:
10/Pk.
Formato:
90mm Monoplate
Tipo di media:
CNA Agar with Sheep Blood
Include:
Sheep Blood
Dimensione della confezione:
pezzi 10
Descrizione
Effectively isolate Gram-positive organisms from clinical samples using Thermo Scientific™ Columbia CNA Agar with Sheep Blood, a selective microbiological medium that includes colistin and nalidixic acid to inhibit gram-negative bacilli. Columbia CNA Agar has traditionally been used to identify staphylococci and streptococci. The addition of sheep blood to the medium allows distinct identification of S. pneumoniae through the production of clear alpha-haemolysis.
Columbia CNA Agar with Sheep Blood contains antibiotics to inhibit S. albus and Micrococcus species as well as Gram-positive and Gram-negative rods. It suppresses growth of Proteus, Klebsiella and Pseudomonas species while permitting unrestricted growth of S. aureus, haemolytic streptococci and enterococci. This medium enables important Gram-positive cocci to be recognised more readily and isolated easily from the mixed bacterial populations contained in many clinical specimens and foods.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifica
Specifica
| Colore | Traffic red, opaque |
| Content And Storage | 2° to 12°C, in the dark |
| Formato | 90mm Monoplate |
| Manufacturing Origin | Germany |
| Tipo di media | CNA Agar with Sheep Blood |
| Product Type | Agar |
| Peso riempimento | 17g ± 5% |
| Include | Sheep Blood |
| Tipo di imballaggio | Flow Wrapped - Single Layer |
| Quantità | 10/Pk. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto