Learn More
BRAND™ Combi-Pack Caps
Combi-Pack Caps constructed of polypropylene are equipped with a polyethylene seal.
Marca: BRAND™ 702883
Additional Details : Peso : 0.01000kg
Description
- Seal included
- For use with Transferpettor
Specifications
Polypropylene | |
100 to 500 μL | |
PE Seal |
Cap | |
100 to 500 μL Transferpettor | |
2 x Caps, 1 x Seal |