Learn More
Complement C3 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marca: Invitrogen PA129715
Dettagli aggiuntivi : Peso : 1.24740kg
Descrizione
Specificity: Monospecific to human C3 complement component. This antibody has also been reported to recognize the C3 fragments, C3a, C3b and C3c. Product is supplied as diluted unpurified antiserum. Store as a concentrated solution. Centrifuge briefly prior to opening the vial.
C3 is a key component of the complement system since classical and alternative activation pathways merge at the C3 activation step when C3 is split into C3a and C3b. The molecular mass of C3 is 185 kDa and it consists of two chains (110 kDa and 75 kDa) held together by disulfide bonds.Specifica
Complement C3 | |
Polyclonal | |
Unconjugated | |
C3 | |
Acylation stimulating protein; acylation-stimulating protein cleavage product; AHUS5; AI255234; ARMD9; ASP; C3; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1; C3a; C3a anaphylatoxin; C3adesArg; C3b; C3bc; C3-beta-c; C3d; complement C3; Complement C3 alpha chain; Complement C3 beta chain; complement C3 protein (GPC3) precursor; Complement C3b alpha' chain; Complement C3c alpha' chain fragment 1; Complement C3c alpha' chain fragment 2; Complement C3d fragment; Complement C3dg fragment; Complement C3f fragment; Complement C3g fragment; complement component 3; complement component C3; complement component C3 alpha-chain; complement component C3a; complement component C3b; complement component C3d; CPAMD1; ENCF-1; ENCF-2; epididymis secretory sperm binding protein Li 62p; HEL-S-62p; HSE-MSF; I79_008227; LOC100060539; Neutrophil chemotactic factor-1; Neutrophil chemotactic factor-2; Plp; prepro-C3; Unknown (protein for MGC:134562) | |
Goat | |
Purified | |
RUO | |
100009121, 100060539, 100135462, 100763958, 12266, 24232, 397072, 718 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunoelectrophoresis, Western Blot | |
94 mg/mL | |
0.05M tris HCl with 500mM NaCl and 0.1% sodium azide; pH 7.5 | |
P01024, P01025, P01026, P01027, P12247, P12387 | |
C3, LOC100060539 | |
Human complement C3 | |
1 mL | |
Primary | |
Baboon, Equine, Guinea Pig, Hamster, Human, Mouse, Non-human Primate, Porcine, Rabbit, Rat | |
Antibody | |
IgG |