missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard 84μS/cm, Hanna Instruments™
Click to view available options
Quantità:
500 mL
Imballaggio:
FDA Bottle
Light Block Bottles
Specifica
Specifica
Imballaggio | Light Block Bottles |
Quantità | 500 mL |
Colore | Colorless |
Odore | Odorless |
pH | 7 |
Solubility | Soluble |
Forma fisica | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'