Learn More
Corning™ PureCoat™ Amine Flat Bottom Multiwell Plates
Corning PureCoat Amine (positively charged) and Carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.
Marca: Corning™ 354721
Dettagli aggiuntivi : Peso : 0.46000kg
Descrizione
Corning PureCoat Amine (positively charged) and Carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.
Specifica
2 x 3 | |
Polystyrene | |
Sterile | |
Multi-well Cell Culture Plate | |
Culture of primary cells, transfected, transformed and fastidious cell lines, culture in standard, serum-free or serum-reduced conditions, freeze-thaw recovery of cryopreserved primary cells and cell lines, including division arrested | |
5/Pk. |
Flat | |
Rectangular | |
Amine | |
6 | |
5/Cs. | |
5/Cs. |