Learn More
Hirschmann™ Coperchi in vetro
Codice prodotto. 10531942
Click to view available options
Dimensioni (L x P):
15 x 15 mm
18 x 18 mm
20 x 20 mm
22 x 22 mm
22 x 32 mm
22 x 40 mm
22 x 50 mm
22 x 60 mm
24 x 24 mm
24 x 32 mm
24 x 46 mm
24 x 48 mm
24 x 50 mm
Dimensione della confezione:
pezzi 1000
Codice prodotto. 10531942
Marca: Hirschmann™ 8000103
Preserve and protect specimens on slides. Hirschmann Instruments™ Cover Glasses for Microscopy are manufactured from quality glass and come in a range of dimensions.
  • 0.13-0.17 mm thickness
  • Acc. to DIN 58 884, CE acc. IVD 98/79 EC
Specifica
Product Type Coverslip
Spessore (sistema metrico) #1 (0.13 to 0.16 mm)
Dimensioni (L x P) 20 x 20 mm
Certifications CE, DIN 58884, IVD 98/79 EC